Merck
  • Development of poly (I:C) modified doxorubicin loaded magnetic dendrimer nanoparticles for targeted combination therapy.
Development of poly (I:C) modified doxorubicin loaded magnetic dendrimer nanoparticles for targeted combination therapy.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2014-12-03)
Rouhollah Khodadust, Gozde Unsoy, Ufuk Gunduz
ABSTRACT
The objective of this study was to develop and evaluate the anticancer activity and the safety of a combinational drug delivery system using polyamidoamine (PAMAM) dendrimer-coated iron oxide nanoparticles for doxorubicin and poly I:C delivery in vitro. Dendrimer-coated magnetic nanoparticles (DcMNPs) are suitable for drug delivery system as nanocarriers with their following properties, such as surface functional groups, symmetry perfection, internal cavities, nano-size and magnetization. These nanoparticles could be targeted to the tumor site under a magnetic field since they have a magnetic core. DcMNPs were found as a convenient vehicle for targeted doxorubicin delivery in cancer therapy. Poly (I:C) binding on doxorubicin loaded DcMNPs (DcMNPs-Dox) was reported for the first time in the literature. It was also demonstrated that loading of doxorubicin into the cavities of DcMNPs increases the binding efficiency of poly (I:C) to the surface functional groups of dendrimer up to 10 times. When we compare the in vitro cytotoxic properties of doxorubicin, poly (I:C) and poly (I:C) bound doxorubicin loaded DcMNPs (PIC-DcMNPs-Dox), it was observed that PIC-DcMNPs-Dox show the highest cytotoxic effect by passing the cell resistance mechanisms on doxorubicin resistant MCF7 (MCF7/Dox) cells. Results demonstrated that applying PIC-DcMNPs-Dox would improve the efficacy by increasing the biocompatibility of system in blood stream and the toxicity inside tumor cells. These results provide invaluable information and new insight for the design and optimization of a novel combinational drug delivery system for targeted cancer therapy.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Metanolo, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Acido acetico, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acido acetico, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Metanolo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Carbonato di sodio, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Carbonato di sodio, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Metanolo, anhydrous, 99.8%
Sigma-Aldrich
Acido acetico, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Carbonato di sodio, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Acido acetico, suitable for HPLC
Sigma-Aldrich
Acido acetico, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acido acetico, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Metanolo, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
N-(3-dimetilaminopropil)-N′-etilcarbodiimide, commercial grade, powder
Sigma-Aldrich
Metanolo, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Carbonato di sodio, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Etanolo, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Metanolo, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Etanolo, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
1-Methylimidazole, ReagentPlus®, 99%
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%