Merck
  • TiO2 nanoparticles tested in a novel screening whole human blood model of toxicity trigger adverse activation of the kallikrein system at low concentrations.
TiO2 nanoparticles tested in a novel screening whole human blood model of toxicity trigger adverse activation of the kallikrein system at low concentrations.
Biomaterials (2015-03-17)
Barbro Ekstrand-Hammarström, Jaan Hong, Padideh Davoodpour, Kerstin Sandholm, Kristina N Ekdahl, Anders Bucht, Bo Nilsson
ABSTRACT
There is a compelling need to understand and assess the toxicity of industrially produced nanoparticles (NPs). In order to appreciate the long-term effects of NPs, sensitive human-based screening tests that comprehensively map the NP properties are needed to detect possible toxic mechanisms. Animal models can only be used in a limited number of test applications and are subject to ethical concerns, and the interpretation of experiments in animals is also distorted by the species differences. Here, we present a novel easy-to-perform highly sensitive whole-blood model using fresh non-anticoagulated human blood, which most justly reflects complex biological cross talks in a human system. As a demonstrator of the tests versatility, we evaluated the toxicity of TiO2 NPs that are widely used in various applications and otherwise considered to have relatively low toxic properties. We show that TiO2 NPs at very low concentrations (50 ng/mL) induce strong activation of the contact system, which in this model elicits thromboinflammation. These data are in line with the finding of components of the contact system in the protein corona of the TiO2 NPs after exposure to blood. The contact system activation may lead to both thrombotic reactions and generation of bradykinin, thereby representing fuel for chronic inflammation in vivo and potentially long-term risk of autoimmunity, arteriosclerosis and cancer. These results support the notion that this novel whole-blood model represents an important contribution to testing of NP toxicity.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
3,3′-diaminobenzidina, tablet, 10 mg substrate per tablet
Sigma-Aldrich
Ellagic acid, ≥95% (HPLC), powder, from tree bark
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Supelco
Urea, analytical standard
Sigma-Aldrich
Urea, meets USP testing specifications
Supelco
Ellagic acid, analytical standard
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C