Merck
The inhibition effects of insulin on BMP2-induced muscle heterotopic ossification.
Biomaterials (2014-08-19)
Jing Zhang, Yannan Zhao, Xianglin Hou, Bing Chen, Zhifeng Xiao, Jin Han, Chunying Shi, Jianzhou Liu, Qi Miao, Jianwu Dai
ABSTRACT
Bone morphogenetic proteins (BMPs) play an important role in regulating osteoblastic differentiation and bone formation. But the diffuse of BMPs into muscle tissues around bone injury sites often leads to heterotopic ossification, which has been regarded as one of major side-effects of BMP implementation in bone defect patients. It raises great demands for exploring effective methods that preventing BMP-induced heterotopic ossification while not interrupting the osteoinductive activity of BMPs for in situ bone defect repair. Here we found insulin, a positive regulator for bone regeneration, inhibited BMP2-induced muscle heterotopic ossification by suppressing the expression of bone transcription factor Osterix. By analyzing downstream molecules of insulin pathway, we found AKT/mTOR/GSK3 signaling was responsible for the inhibition of insulin on BMP2-induced ossification, and GSK3 inhibitor SB216763 attenuated BMP2-induced muscle heterotopic ossification. The data might shed light on developing effective clinical therapy for inhibiting muscle heterotopic ossification when BMPs were used bone defect repair.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
L-glutammina, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
L-glutammina, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
SAFC
L-glutammina
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Supelco
Electrolyte solution, nonaqueous, 2 M LiCl in ethanol
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
L-glutammina, BioUltra, ≥99.5% (NT)
Supelco
Lithium chloride solution, 1 M in ethanol
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Supelco
Electrolyte solution, nonaqueous, LiCl in ethanol (saturated)
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
L-glutammina, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
L-glutammina
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Lithium chloride, puriss. p.a., anhydrous, ≥99.0% (AT)
Supelco
L-glutammina, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis