Merck
  • Thioether analogues of disulfide-bridged cyclic peptides targeting death receptor 5: conformational analysis, dimerisation and consequences for receptor activation.
Thioether analogues of disulfide-bridged cyclic peptides targeting death receptor 5: conformational analysis, dimerisation and consequences for receptor activation.
Chembiochem : a European journal of chemical biology (2014-12-10)
Karolina Pulka-Ziach, Valeria Pavet, Neila Chekkat, Karine Estieu-Gionnet, Roman Rohac, Marie-Charlotte Lechner, Cristian R Smulski, Gabrielle Zeder-Lutz, Danièle Altschuh, Hinrich Gronemeyer, Sylvie Fournel, Benoit Odaert, Gilles Guichard
ABSTRACT
Cyclic peptides containing redox-stable thioether bridges might provide a useful alternative to disulfide-bridged bioactive peptides. We report the effect of replacing the disulfide bridge with a lanthionine linkage in a 16-mer cyclic peptide that binds to death receptor 5 (DR5, TRAIL-R2). Upon covalent oligomerisation, the disulfide-bridged peptide has previously shown similar behaviour to that of TNF-related apoptosis inducing ligand (TRAIL), by selectively triggering the DR5 cell death pathway. The structural and biological properties of the DR5-binding peptide and its desulfurised analogue were compared. Surface plasmon resonance (SPR) data suggest that these peptides bind DR5 with comparable affinities. The same holds true for dimeric versions of these peptides: the thioether is able to induce DR5-mediated apoptosis of BJAB lymphoma and tumorigenic BJELR cells, albeit to a slightly lower extent compared to its disulfide homologue. NMR analysis revealed subtle variation in the conformations of the two peptides and suggests that the thioether peptide is slightly less folded than its disulfide homologue. These observations could account for the different capability of the two dimers to cluster DR5 receptors on the cell surface and to trigger apoptosis. Nevertheless, our results suggest that the thioether peptide is a potential candidate for evaluation in animal models.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acqua, suitable for HPLC
Sigma-Aldrich
Acido trifluoroacetico, ReagentPlus®, 99%
Sigma-Aldrich
Acido trifluoroacetico, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acqua, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Acqua, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Acido trifluoroacetico, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Acqua, HPLC Plus
Sigma-Aldrich
Acqua, Deionized
Sigma-Aldrich
Ioduro di propidio, ≥94.0% (HPLC)
Supelco
Acqua, suitable for ion chromatography
Sigma-Aldrich
Acido trifluoroacetico, ≥99%, for protein sequencing
Sigma-Aldrich
Acqua, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Acqua, Molecular Biology, sterile filtered
Supelco
Acqua, for TOC analysis
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Acqua, BioPerformance Certified
Supelco
Acido trifluoroacetico, analytical standard
Acqua, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Acqua, ACS reagent
Sigma-Aldrich
Ioduro di propidio, ≥94% (HPLC)
Supelco
Acqua, for ultratrace analysis, ACS reagent
Sigma-Aldrich
Acqua, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Acqua, PCR Reagent
Sigma-Aldrich
Acqua, tested according to Ph. Eur.
Sigma-Aldrich
Acido trifluoroacetico, ReagentPlus®, 99%
Supelco
Acqua, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Acqua, endotoxin, free
Acqua, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Acqua, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O