Merck
Dynamics and cellular localization of Bmp2, Bmp4, and Noggin transcription in the postnatal mouse skeleton.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research (2014-07-22)
Steven K Pregizer, Douglas P Mortlock
ABSTRACT
Transcription of BMPs and their antagonists in precise spatiotemporal patterns is essential for proper skeletal development, maturation, maintenance, and repair. Nevertheless, transcriptional activity of these molecules in skeletal tissues beyond embryogenesis has not been well characterized. In this study, we used several transgenic reporter mouse lines to define the transcriptional activity of two potent BMP ligands, Bmp2 and Bmp4, and their antagonist, Noggin, in the postnatal skeleton. At 3 to 4 weeks of age, Bmp4 and Noggin reporter activity was readily apparent in most cells of the osteogenic or chondrogenic lineages, respectively, whereas Bmp2 reporter activity was strongest in terminally differentiated cells of both lineages. By 5 to 6 months, activity of the reporters had generally abated; however, the Noggin and Bmp2 reporters remained remarkably active in articular chondrocytes and persisted there indefinitely. We further found that endogenous Bmp2, Bmp4, and Noggin transcript levels in postnatal bone and cartilage mirrored the activity of their respective reporters in these tissues. Finally, we found that the activity of the Bmp2, Bmp4, and Noggin reporters in bone and cartilage at 3 to 4 weeks could be recapitulated in both osteogenic and chondrogenic culture models. These results reveal that Bmp2, Bmp4, and Noggin transcription persists to varying degrees in skeletal tissues postnatally, with each gene exhibiting its own cell type-specific pattern of activity. Illuminating these patterns and their dynamics will guide future studies aimed at elucidating both the causes and consequences of aberrant BMP signaling in the postnatal skeleton.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Cloroformio, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Cloroformio, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Cloroformio, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Cloroformio, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
D-(+)-Glucosio, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucosio, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucosio, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Glucosio, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Cloroformio, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Cloroformio, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Supelco
D-(+)-Glucosio, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cloroformio, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Cloroformio, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Cloroformio, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Cloroformio, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
USP
Destrosio, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cloroformio, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
D-(+)-Glucosio, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Cloroformio, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
D-(+)-Glucosio, ACS reagent
Supelco
D-(+)-Glucosio, analytical standard
Sigma-Aldrich
Cloroformio, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Cloroformio, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
D-(+)-Glucosio, suitable for mouse embryo cell culture, ≥99.5% (GC)
Supelco
Cloroformio, analytical standard
Supelco
Cloroformio, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Glucosio, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
Cloroformio, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Cloroformio, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Glucosio, tested according to Ph. Eur.