Merck
  • Potato chip intake increases ascorbic acid levels and decreases reactive oxygen species in SMP30/GNL knockout mouse tissues.
Potato chip intake increases ascorbic acid levels and decreases reactive oxygen species in SMP30/GNL knockout mouse tissues.
Journal of agricultural and food chemistry (2014-09-03)
Yoshitaka Kondo, Rui Sakuma, Megumi Ichisawa, Katsuyuki Ishihara, Misako Kubo, Setsuko Handa, Hiroyuki Mugita, Naoki Maruyama, Hidenori Koga, Akihito Ishigami
ABSTRACT
Potato chips (PC) contain abundant amounts of the free radical scavenger ascorbic acid (AA) due to the rapid dehydration of potato tubers (Solanum tuberosum) that occurs during frying. To evaluate the antioxidant activity of PC, this study examined reactive oxygen species (ROS) levels in tissues from SMP30/GNL knockout (KO) mice that cannot synthesize AA and determined AA and ROS levels after the animals were fed 20 and 10% PC diets for 7 weeks. Compared with AA-sufficient mice, AA-depleted SMP30/GNL KO mice showed high ROS levels in tissues. SMP30/GNL KO mice fed a PC diet showed high AA and low ROS levels in the brain, heart, lung, testis, soleus muscle, plantaris muscle, stomach, small intestine, large intestine, eyeball, and epididymal fat compared with AA-depleted mice. The data suggest that PC intake increases AA levels and enhances ROS scavenging activity in tissues of SMP30/GNL KO mice, which are a promising model for evaluating the antioxidant activity of foods.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acido L-ascorbico, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Acido L-ascorbico, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Acido L-ascorbico, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Acido L-ascorbico, 99%
Sigma-Aldrich
Acido L-ascorbico, reagent grade, crystalline
Supelco
Acido L-ascorbico, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
USP
Acido ascorbico, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Acido L-ascorbico, ACS reagent, ≥99%
Supelco
Acido L-ascorbico, analytical standard
Sigma-Aldrich
Acido L-ascorbico, meets USP testing specifications
Sigma-Aldrich
Acido L-ascorbico, reagent grade
Sigma-Aldrich
Acido L-ascorbico, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Acido L-ascorbico, FCC, FG
Sigma-Aldrich
Acido L-ascorbico, BioUltra, ≥99.5% (RT)
Acido L-ascorbico, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Acido L-ascorbico, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
Acido L-ascorbico, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acido L-ascorbico, tested according to Ph. Eur.