Merck
Polybutylene terephthalate-nickel oxide nanocomposite as a fiber coating.
Analytica chimica acta (2015-03-04)
Habib Bagheri, Ali Roostaie
ABSTRACT
A highly efficient polybutylene terephthalate (PBT)-based nanocomposite containing nickel oxide nanoparticles was synthesized by electrospinning technique and used as a fiber coating for solid phase microextraction. The influential morphological parameters and capability of the prepared nanocomposite including the NiO content, the coating time, the PBT concentration and applied voltage were considered for optimization. The applicability of the synthesized fiber coating was examined by headspace solid phase micro extraction and gas chromatography mass spectrometry detection of some volatile organic compounds in aqueous samples. Among the synthesized nanocomposites and pristine PBT nanofibers, the fiber coating with 14% of NiO nanoparticles doping level exhibited the highest extraction efficiency. In addition, important parameters influencing the extraction/desorption process were investigated and optimized. The detection limits were less than 5 ng L(-1) using the selected ion monitoring mode. The inter-day and intra-day precisions of the developed method under optimized conditions were below 11%. The method showed a linearity in the range of 10-1000 ng L(-1) with the correlation coefficient greater than 0.9987. The optimized method was applied to the sampling of some volatile organic compounds from real water samples. The developed method is convenient, rapid, simple, easy and inexpensive and offers high sensitivity and sufficient reproducibility.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Metanolo, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Metanolo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Metanolo, anhydrous, 99.8%
Sigma-Aldrich
Sodio cloruro, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Sodio cloruro, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodio cloruro, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodio cloruro, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodio cloruro, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Metanolo, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Metanolo, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Metanolo, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
SAFC
Sodio cloruro, 5 M
Supelco
Gruppo in fibra SPME divinilbenzene/carbossene/polidimetilsilossano (DVB/CAR/PDMS), 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Supelco
Gruppo in fibra SPME divinilbenzene/carbossene/polidimetilsilossano (DVB/CAR/PDMS), 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub
Supelco
Gruppo in fibra SPME divinilbenzene/carbossene/polidimetilsilossano (DVB/CAR/PDMS), 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Sigma-Aldrich
Metanolo, Absolute - Acetone free
USP
Metanolo, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Metanolo, BioReagent, ≥99.93%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%