Merck
  • Ultraviolet-C irradiation to titanium implants increases peri-implant bone formation without impeding mineralization in a rabbit femur model.
Ultraviolet-C irradiation to titanium implants increases peri-implant bone formation without impeding mineralization in a rabbit femur model.
Acta odontologica Scandinavica (2015-02-04)
Makoto Yamazaki, Masahiro Yamada, Ken Ishizaki, Kaoru Sakurai
ABSTRACT
Volume and bone quality of peri-implant supporting bone, in particular, at implant neck region, as well as bone-implant contact ratio, is important for long-term stability of implants. Ultraviolet-C (UVC) irradiation is known to enhance the osseointegration capability of titanium implants. However, the histological determination was performed only on a rat model, but not pre-clinical animal model such as a rabbit model. The purpose of this study was to determine the effects of UVC irradiation on titanium implants on the volume and mineral density of peri-implant supporting bone formation in a rabbit femur model. Acid-etched pure titanium screw implants with or without 3 mW/cm2 UVC irradiation for 48 h were placed in rabbit femur diaphyses. Peri-implant bone tissue formation was analyzed at 3 and 8 weeks post-operatively by histology and micro-CT-based bone morphometry after calibration with hydroxyl apatite phantoms. UVC pre-irradiated implants accumulated a higher density of cells and thicker and longer bone tissue attachments that continued into the inner basic lamellae of the surface of existing cortical bone at 3 and 8 weeks than the implants without irradiation. Although the bone mineral density around both implants was equivalent to that of the existing cortical bone, bone volume was greater with UVC pre-irradiation in two-thirds or more of the apical region throughout the observation period. These results indicate that UVC treatment increased the volume of cortical-like bone tissue in the coronal region of titanium implants without deterioration of bone mineral density.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Lidocaine, powder
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Lidocaine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Lidocaine, analytical standard
Adrenaline, European Pharmacopoeia (EP) Reference Standard
Lidocaine, European Pharmacopoeia (EP) Reference Standard