Merck
Genome-wide association study identifies a new locus for coronary artery disease on chromosome 10p11.23.
European heart journal (2010-11-23)
Jeanette Erdmann, Christina Willenborg, Janja Nahrstaedt, Michael Preuss, Inke R König, Jens Baumert, Patrick Linsel-Nitschke, Christian Gieger, Stephanie Tennstedt, Petra Belcredi, Zouhair Aherrahrou, Norman Klopp, Christina Loley, Klaus Stark, Christian Hengstenberg, Petra Bruse, Jennifer Freyer, Arnika K Wagner, Anja Medack, Wolfgang Lieb, Anika Grosshennig, Hendrik B Sager, Adrian Reinhardt, Arne Schäfer, Stefan Schreiber, Nour Eddine El Mokhtari, Dorette Raaz-Schrauder, Thomas Illig, Christoph D Garlichs, Arif B Ekici, André Reis, Jürgen Schrezenmeir, Diana Rubin, Andreas Ziegler, H-E Wichmann, Angela Doering, Christa Meisinger, Thomas Meitinger, Annette Peters, Heribert Schunkert
ABSTRACT
Recent genome-wide association (GWA) studies identified 10 chromosomal loci for coronary artery disease (CAD) or myocardial infarction (MI). However, these loci explain only a small proportion of the genetic variability of these pertinent diseases. We sought to identify additional CAD/MI loci by applying a three-stage approach. We genotyped n = 1157 MI cases and n = 1748 controls from a population-based study population [German MI Family Study (GerMIFS) III (KORA)] with genome-wide SNP arrays. At this first stage, n = 462 SNPs showed association with MI at P<1 × 10(-3) in two-sided logistic regression. In a second stage, 415 of these SNPs were evaluated in silico in two independent GWA samples, the GerMIFS I (875 cases/1644 controls) and GerMIFS II (1222 cases/1298 controls). Nine SNPs, representing three regions, displayed consistent replication in this in silico analysis (P<0.05 for each GWA sample): five SNPs at 9p21.3, a well-known CAD/MI locus, two SNPs at 10p11.21, and two SNPs at 2p24.3. Wet-lab replication, i.e. the third stage, of SNP rs3739998 (representing the novel locus at 10p11.21, p.S1002T in the KIAA1462 gene) in additional 5790 cases and 5302 controls confirmed the association (P=9.54 × 10(-4)), but not for the 2p24.3 locus. The combined P-value across all stages for SNP rs3739998 is P=1.27 × 10(-11) [odds ratio (OR) = 1.15 (1.11-1.20)]. Analysis of a GWA study followed by in silico and wet-lab replication steps identified the KIAA1462 gene, encoding a yet uncharacterized protein, on chromosome 10p11.23 with genome-wide significant association for CAD/MI. Further studies are needed to characterize the functional role of this locus in the aetiology of these diseases.