Merck
  • Noggin-Mediated Retinal Induction Reveals a Novel Interplay Between Bone Morphogenetic Protein Inhibition, Transforming Growth Factor β, and Sonic Hedgehog Signaling.
Noggin-Mediated Retinal Induction Reveals a Novel Interplay Between Bone Morphogenetic Protein Inhibition, Transforming Growth Factor β, and Sonic Hedgehog Signaling.
Stem cells (Dayton, Ohio) (2015-04-29)
Andrea Messina, Lei Lan, Tania Incitti, Angela Bozza, Massimiliano Andreazzoli, Robert Vignali, Federico Cremisi, Yuri Bozzi, Simona Casarosa
ABSTRACT
It has long been known that the depletion of bone morphogenetic protein (BMP) is one of the key factors necessary for the development of anterior neuroectodermal structures. However, the precise molecular mechanisms that underlie forebrain regionalization are still not completely understood. Here, we show that Noggin1 is involved in the regionalization of anterior neural structures in a dose-dependent manner. Low doses of Noggin1 expand prosencephalic territories, while higher doses specify diencephalic and retinal regions at the expense of telencephalic areas. A similar dose-dependent mechanism determines the ability of Noggin1 to convert pluripotent cells in prosencephalic or diencephalic/retinal precursors, as shown by transplant experiments and molecular analyses. At a molecular level, the strong inhibition of BMP signaling exerted by high doses of Noggin1 reinforces the Nodal/transforming growth factor (TGF)β signaling pathway, leading to activation of Gli1 and Gli2 and subsequent activation of Sonic Hedgehog (SHH) signaling. We propose a new role for Noggin1 in determining specific anterior neural structures by the modulation of TGFβ and SHH signaling.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
SB 431542, ≥98% (HPLC), powder
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Anticorpo anti-fosfo-istone H3 (Ser10) marcatore di mitosi, Upstate®, from rabbit
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Phosphorus, red, ≥99.99% trace metals basis
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Phosphorus, red, ≥97.0%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Acido bicinconinico, ≥98% (HPLC)
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
3-Maleimidobenzoic acid N-hydroxysuccinimide ester, crystalline
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Cyclopamine hydrate, ≥98% (HPLC)
Prezzi e disponibilità al momento non sono disponibili