Merck
  • Synthesis of surface nano-molecularly imprinted polymers for sensitive baicalin detection from biological samples.
Synthesis of surface nano-molecularly imprinted polymers for sensitive baicalin detection from biological samples.
RSC advances (2015-08-11)
Xiaoli Gu, Hongliang He, Chong-Zhi Wang, Yankun Gao, Hongjuan Zhang, Junli Hong, Shuhu Du, Lina Chen, Chun-Su Yuan
ABSTRACT
Surface molecularly imprinted polymers (MIP@SBA-15) imprinted on the surface of hybrid nanostructured organic/inorganic materials (SBA-15) were prepared for the selective extraction and detection of baicalin (BA) from biological samples. The surface morphologies and characteristics of the imprinted and non-imprinted polymers were characterized by Fourier transform infrared (FT-IR), scanning electron microscopy (SEM), transmission electron microscopy (TEM), thermo-gravimetric analysis (TGA) and nitrogen adsorption-desorption isotherms. The results indicated that the polymers were successfully grafted on the surface of SBA-15 and possessed a highly ordered mesoporous structure. In binding tests, MIP@SBA-15 reached saturated adsorption within 80 min and exhibited significant specific recognition toward BA with large adsorption capacity. Meanwhile, the prepared MIP@SBA-15 was used as a selective sorbent for solid-phase extraction of BA from biological samples. Recoveries of BA from the liver and spleen ranged from 90.6% to 90.9% with RSD < 3.7%. All these results reveal that this method is simple, rapid and sensitive for effectively extracting and detecting trace BA in biological samples.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
tetra-etil ortosilicato, reagent grade, 98%
Sigma-Aldrich
Metanolo, anhydrous, 99.8%
Sigma-Aldrich
Acido cloridrico, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
tetra-etil ortosilicato, ≥99.0% (GC)
Sigma-Aldrich
Etilenglicole dimetilacrilato, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Acido cloridrico, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Acido cloridrico, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
4-Vinylpyridine, contains 100 ppm hydroquinone as inhibitor, 95%
Sigma-Aldrich
Acido cloridrico, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
tetra-etil ortosilicato, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Cloruro di idrogeno, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Acido cloridrico, 32 wt. % in H2O, FCC
Sigma-Aldrich
Flavone, ≥99.0%
Sigma-Aldrich
tetra-etil ortosilicato, packaged for use in deposition systems
Sigma-Aldrich
Metanolo, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.