Merck
  • Activin receptor-like kinases: a novel subclass of cell-surface receptors with predicted serine/threonine kinase activity.
Activin receptor-like kinases: a novel subclass of cell-surface receptors with predicted serine/threonine kinase activity.
Oncogene (1993-10-01)
P ten Dijke, H Ichijo, P Franzén, P Schulz, J Saras, H Toyoshima, C H Heldin, K Miyazono
ABSTRACT
Human cDNA clones encoding four novel putative transmembrane protein serine/threonine kinases, denoted activin receptor-like kinase (ALK) -1, -2, -3 and -4, were obtained using a polymerase chain reaction (PCR)-based strategy. The PCR primers were designed based upon the sequence similarity between the activin receptor type II and Daf-1. The cDNA clones for ALK-1, -2 and -3 encode complete proteins of 503, 509 and 532 amino acids respectively. The ALK-4 cDNA is incomplete and the predicted protein of 383 amino acids has a truncated extracellular domain. The ALKs share similar domain structures, comprising predicted signal sequences at the N-terminals, followed by hydrophilic cysteine-rich ligand-binding domains, single hydrophobic transmembrane regions and C-terminal intracellular portions that consist almost entirely of putative serine/threonine kinase domains. The ALKs have approximately 40% sequence identity to activin receptors type II and IIB, transforming growth factor-beta (TGF-beta) type II receptor and Daf-1 in the kinase domains. However, the sequence identities are higher (60-79%) between ALK-1, -2, -3 and -4, suggesting that they form a subfamily among the putative receptor serine/threonine kinases. The extracellular domains of ALKs show only little sequence identity to other putative receptor serine/threonine kinases, but the cysteine residues are conserved. Their structural properties suggest that ALK-1 to -4 are receptors that may bind ligands that are members of the TGF-beta superfamily. The expression of mRNA in human tissues varied for the different ALKs; ALK-2 and ALK-4 showed ubiquitous tissue expression patterns, whereas the distribution of ALK-1 and ALK-3 varied strongly between different tissues with more restricted expression patterns. These results suggest that each ALK may have different in vivo functions.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Activin Receptor-Like Kinase 1/Fc Chimera human, >90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture