Merck
Short- and long-term regulation of intestinal Na+/H+ exchange by Toll-like receptors TLR4 and TLR5.
American journal of physiology. Gastrointestinal and liver physiology (2015-08-22)
José Miguel Cabral, Daniela Grácio, Patrício Soares-da-Silva, Fernando Magro
ABSTRACT
Inappropriate activation of pattern recognition receptors has been described as a potential trigger in the development of inflammatory bowel disease (IBD). In this study, we evaluated the activity and expression of Na(+)/H(+) exchanger (NHE) subtypes in T84 intestinal epithelial cells during Toll-like receptor 4 (TLR4) activation by monophosphoryl lipid A and TLR5 by flagellin. NHE activity and intracellular pH were evaluated by spectrofluorescence. Additionally, kinase activities were evaluated by ELISA, and siRNA was used to specifically inhibit adenylyl cyclase (AC). Monophosphoryl lipid A (MPLA) (0.01-50.00 μg/ml) and flagellin (10-500 ng/ml) inhibited NHE1 activity in a concentration-dependent manner (MPLA short term -25.2 ± 5.0%, long term -31.9 ± 4.0%; flagellin short term -14.9 ± 2.0%, long term -19.1 ± 2.0%). Both ligands triggered AC3, PKA, PLC, and PKC signal molecules. Long-term exposure to flagellin and MPLA induced opposite changes on NHE3 activity; flagellin increased NHE3 activity (∼10%) with overexpression of membrane protein, whereas MPLA decreased NHE3 activity (-17.3 ± 3.0%). MPLA and flagellin simultaneously had synergistic effects on NHE activity. MPLA and flagellin impaired pHi recovery after intracellular acidification. The simultaneous exposure to MPLA and flagellin induced a substantial pHi reduction (-0.55 ± 0.03 pH units). Activation of TLR4 and TLR5 exerts marked inhibition of NHE1 activity in intestinal epithelial cells. Transduction mechanisms set into motion during TLR4-mediated and long-term TLR5-mediated inhibition of NHE1 activity involve AC3, PKA, PLC, and PKC. However, short- and long-term TLR4 activation and TLR5 activation might use different signaling pathways. The physiological alterations on intestinal epithelial cells described here may be useful in the development of better IBD therapeutics.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodio cloruro, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodio cloruro, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodio cloruro, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Cloruro di calcio, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodio cloruro, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodio cloruro, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Trietanolammina, ≥99.0% (GC)
Sigma-Aldrich
Forskolina, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Forskolina, For use in molecular biology applications
Sigma-Aldrich
Cloruro di calcio, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
SAFC
Sodio cloruro, 5 M
Sigma-Aldrich
Soluzione tampone HEPES, 1 M in H2O
Sigma-Aldrich
Trietanolammina, reagent grade, 98%
Sigma-Aldrich
Sodio cloruro, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodio cloruro, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Cloruro di calcio, powder, 99.99% trace metals basis
Sigma-Aldrich
Cloruro di calcio
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Cloruro di calcio, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Trietanolammina, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodio cloruro, 0.85%
Sigma-Aldrich
Trietanolammina, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Trietanolammina, puriss., meets analytical specification of NF, ≥99% (GC)
Sigma-Aldrich
Phorbol 12,13-dibutyrate, ≥98% (TLC), powder
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl