Merck
  • Cholinesterase-inhibitory diterpenoids and chemical constituents from aerial parts of Caryopteris mongolica.
Cholinesterase-inhibitory diterpenoids and chemical constituents from aerial parts of Caryopteris mongolica.
Journal of natural medicines (2015-04-23)
Toshihiro Murata, Erdenechimeg Selenge, Saki Oikawa, Keita Ageishi, Javzan Batkhuu, Kenroh Sasaki, Fumihiko Yoshizaki
ABSTRACT
A diterpenoid diglucoside (12,19-di-O-β-D-glucopyranosyl-11-hydroxyabieta-8,11,13-triene-19-one), isoscutellarein 7-O-[β-D-xylopyranosyl-(1→2)]-β-D-glucopyranoside, isoscutellarein 7-O-[α-L-rhamnopyranosyl-(1→2)]-β-D-glucopyranoside, hypolaetin 7-O-[6″-O-(p-E-coumaroyl)]-β-D-glucopyranoside, hypolaetin 7-O-[6″-O-(E-caffeoyl)]-β-D-glucopyranoside, and 15 known compounds were isolated from aerial parts of the Mongolian medicinal plant Caryopteris mongolica. The cholinesterase-inhibitory activities of the constituents were estimated. The abietane diterpenoids (12-O-demethylcryptojaponol and 6α-hydroxydemethylcryptojaponol) showed potent inhibitory activity against acetylcholinesterase from human erythrocytes and electric eel, and against butyrylcholinesterase from horse serum.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Glicerolo, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glicerolo, ≥99.5%
Sigma-Aldrich
Glicerolo, 83.5-89.5% (T)
Sigma-Aldrich
Glicerolo, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glicerolo, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glicerolo, BioXtra, ≥99% (GC)
Sigma-Aldrich
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
Sigma-Aldrich
Glicerolo, FCC, FG
Sigma-Aldrich
Glicerolo, meets USP testing specifications
Sigma-Aldrich
Acetylthiocholine iodide, ≥99.0% (AT)
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
Glicerolo, Vetec, reagent grade, 99%