Merck
Differential Microscopic Mobility of Components within a Deep Eutectic Solvent.
The journal of physical chemistry letters (2015-08-13)
Durgesh V Wagle, Gary A Baker, Eugene Mamontov
ABSTRACT
From macroscopic measurements of deep eutectic solvents such as glyceline (1:2 molar ratio of choline chloride to glycerol), the long-range translational diffusion of the larger cation (choline) is known to be slower compared to that of the smaller hydrogen bond donor (glycerol). However, when the diffusion dynamics are analyzed on the subnanometer length scale, we find that the displacements associated with the localized diffusive motions are actually larger for choline. This counterintuitive diffusive behavior can be understood as follows. The localized diffusive motions confined in the transient cage of neighbor particles, which precede the cage-breaking long-range diffusion jumps, are more spatially constrained for glycerol than for choline because of the stronger hydrogen bonds the former makes with chloride anions. The implications of such differential localized mobility of the constituents should be especially important for applications where deep eutectic solvents are confined on the nanometer length scale and their long-range translational diffusion is strongly inhibited (e.g., within microporous media).
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Glicerolo, Molecular Biology, ≥99.0%
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
Glicerolo, ≥99.5%
Sigma-Aldrich
Glicerolo, 83.5-89.5% (T)
Sigma-Aldrich
Glicerolo, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glicerolo, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Glicerolo, BioXtra, ≥99% (GC)
Sigma-Aldrich
Choline chloride, ≥99%
Sigma-Aldrich
Glicerolo, FCC, FG
Sigma-Aldrich
Glicerolo, meets USP testing specifications
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Glicerolo, Vetec, reagent grade, 99%