Merck
Disparate metabolic effects of blackcurrant seed oil in rats fed a basal and obesogenic diet.
European journal of nutrition (2014-10-15)
Adam Jurgoński, Bartosz Fotschki, Jerzy Juśkiewicz
ABSTRACT
It was hypothesised that blackcurrant seed oil beneficially modulates metabolic disorders related to obesity and its complications. The study also aimed to investigate the potentially adverse effects of an unbalanced diet on the distal intestine. Male Wistar rats were randomly assigned to four groups of eight animals each and were fed a basal or obesogenic (high in fat and low in fibre) diet that contained either rapeseed oil (Canola) or blackcurrant seed oil. A two-way analysis of variance was then applied to assess the effects of diet and oil and the interaction between them. After 8 weeks, the obesogenic dietary regimen increased the body weight, altered the plasma lipid profile and increased the liver fat content and the plasma transaminase activities. In addition, the obesogenic diet decreased bacterial glycolytic activity and short-chain fatty acid formation in the distal intestine. Dietary blackcurrant seed oil improved the lipid metabolism by lowering liver fat accumulation and the plasma triglyceride concentration and atherogenicity as well by increasing the plasma HDL-cholesterol concentration. However, in rats fed an obesogenic diet containing blackcurrant seed oil, the plasma HDL-cholesterol concentration was comparable with both rapeseed oil-containing diets, and a significant elevation of the plasma transaminase activities was noted instead. The obesogenic dietary regimen causes a number of metabolic disorders, including alterations in the hindgut microbial metabolism. Dietary blackcurrant seed oil ameliorates the lipid metabolism; however, the beneficial effect is restricted when it is provided together with the obesogenic diet, and a risk of liver injury may occur.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Colesterolo, Sigma Grade, ≥99%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Colesterolo, powder, BioReagent, suitable for cell culture, ≥99%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Colesterolo, from sheep wool, ≥92.5% (GC), powder
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
2-Nitrophenol, 98%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Prezzi e disponibilità al momento non sono disponibili
SAFC
Colesterolo, from sheep wool, Controlled origin, meets USP/NF testing specifications
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Colesterolo, from lanolin, ≥99.0% (GC)
Prezzi e disponibilità al momento non sono disponibili