Merck
  • Crystallization and preliminary X-ray diffraction analysis of a putative carbon-carbon bond hydrolase from Mycobacterium abscessus 103.
Crystallization and preliminary X-ray diffraction analysis of a putative carbon-carbon bond hydrolase from Mycobacterium abscessus 103.
Acta crystallographica. Section F, Structural biology communications (2015-02-11)
Zhang Zhang, Yong-Liang Jiang, Yi Wu, Yong-Xing He
ABSTRACT
The PhlG protein from Mycobacterium abscessus 103 (mPhlG), which shares 30% sequence identity with phloretin hydrolase from Eubacterium ramulus and 38% sequence identity with 2,4-diacetylphloroglucinol hydrolase from Pseudomonas fluorescens Pf-5, is a putative carbon-carbon bond hydrolase. Here, the expression, purification and crystallization of mPhlG are reported. Crystals were obtained using a precipitant consisting of 100 mM citric acid pH 5.0, 1.0 M lithium chloride, 8%(w/v) polyethylene glycol 6000. The crystals diffracted to 1.87 Å resolution and belonged to space group P21, with unit-cell parameters a = 71.0, b = 63.4, c = 74.7 Å, α = 90.0, β = 103.2, γ = 90.0°. Assuming the presence of two mPhlG molecules in the asymmetric unit, VM was calculated to be 2.5 Å(3) Da(-1), which corresponds to a solvent content of 50%.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Glicerolo, Molecular Biology, ≥99.0%
Sigma-Aldrich
2-mercaptoetanolo, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-mercaptoetanolo, ≥99.0%
Sigma-Aldrich
Imidazolo, ReagentPlus®, 99%
Sigma-Aldrich
Imidazolo, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Glicerolo, ≥99.5%
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Glicerolo, 83.5-89.5% (T)
Sigma-Aldrich
Imidazolo, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
2-mercaptoetanolo, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glicerolo, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glicerolo, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Imidazolo, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Glicerolo, BioXtra, ≥99% (GC)
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Imidazolo, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Glicerolo, FCC, FG
Sigma-Aldrich
Glicerolo, meets USP testing specifications
Sigma-Aldrich
Imidazolo, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazolo, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazolo, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Glicerolo, Vetec, reagent grade, 99%
Sigma-Aldrich
Imidazolo, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazolo, Molecular Biology, ≥99% (titration), free-flowing, Redi-Dri