Merck
New Polyphenols Identified in Artemisiae abrotani herba Extract.
Molecules (Basel, Switzerland) (2015-06-18)
Elisabeta Baiceanu, Laurian Vlase, Andrei Baiceanu, Madalina Nanes, Dan Rusu, Gianina Crisan
ABSTRACT
Artemisia abrotanum L. ("southernwood") belongs to the Artemisia genus and it is used in traditional medicine for the treatment of a variety of illnesses. Scarce data is available on the chemical composition of this medicinal plant, most research being focused on the quantitative and qualitative analyses of its essential oil. Our aim was to investigate the content and profile of polyphenols, flavonoids and hydroxycinnamic derivatives present in the Artemisiae abrotani herba extract. We conducted LC/MS analysis and we screened for 19 polyphenols, flavonoids and hydroxycinnamic derivatives. We determined the total content of these compounds and we screened for antioxidant activity. Most polyphenol acids, hydroxycinnamic derivatives and flavonoids were identified and quantified for the first time in this study. We found an original polyphenol distribution profile with high concentration of sinapic acid, rutin, quercetol, ferulic acid and patuletin. We measured the antioxidant activity, the ethanolic extract presenting a modest radical scavenging activity. The value of this study consists in its novelty as it adds new data on the chemical composition of A. abrotanum L. and it opens novel perspectives for medical and nutritional applications of this plant.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Metanolo, anhydrous, 99.8%
Sigma-Aldrich
Idrossido di sodio, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Acido fosforico, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Idrossido di sodio, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acido cloridrico, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acido cloridrico, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Acido caffeico, ≥98.0% (HPLC)
Sigma-Aldrich
Acido fosforico, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Supelco
Acido cloridrico, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
Acido 2,5-diidrossibenzoico, 98%
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Acido fosforico, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Carbonato di sodio, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Carbonato di sodio, BioXtra, ≥99.0%
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Acido cloridrico, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Acido fosforico, BioUltra, ≥85% (T)
Supelco
Acido 2,5-diidrossibenzoico, suitable for matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis