Merck
  • Sevoflurane attenuate hypoxia-induced VEGF level in tongue squamous cell carcinoma cell by upregulating the DNA methylation states of the promoter region.
Sevoflurane attenuate hypoxia-induced VEGF level in tongue squamous cell carcinoma cell by upregulating the DNA methylation states of the promoter region.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2015-05-12)
Yi Lu, Jing Wang, Jia Yan, Yaqiong Yang, Yu Sun, Yan Huang, Rong Hu, Ying Zhang, Hong Jiang
ABSTRACT
Anaesthetic agents were confirmed to play a role on the tumor angiogenesis. The effect of sevoflurane on tongue squamous cell carcinoma (TSCC) cell has not been investigated. SCC-4 cells were exposed to sevoflurane after simulating hypoxia. Then, both the mRNA and protein level of hypoxia-inducible factor (HIF)-1α and VEGF were detected. The methylation states of the VEGF promoter region were also assessed to reveal the underlying mechanism. Finally, the DNA methyltransferase inhibitor 5-aza-2'-deoxycytidine (5-Aza) was administrated to reveal the relationship of DNA methylation on the regulation of the VEGF level. Results showed that sevoflurane attenuated the hypoxia-induced VEGF level without altering the HIF-1α after exposure for 24 and 72 h. Sevoflurane increased the DNA methylation of the VEGF promoter region. The attenuation effect of sevoflurane on hypoxia-induced VEGF level could be blocked by 5-Aza. We concluded that sevoflurane attenuates hypoxia-induced VEGF level via DNA methylation of the promoter region in TSCC cell.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Tiazolil blu tetrazolio bromuro, 98%
Sigma-Aldrich
Tiazolil blu tetrazolio bromuro, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Calceina-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Calceina-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
Hydroquinone, meets USP testing specifications