Merck
  • Functional analysis of differences in transcriptional activity conferred by genetic variants in the 5' flanking region of the IL12RB2 gene.
Functional analysis of differences in transcriptional activity conferred by genetic variants in the 5' flanking region of the IL12RB2 gene.
Immunogenetics (2015-11-11)
Nahoko Kato-Kogoe, Hideki Ohyama, Soichiro Okano, Koji Yamanegi, Naoko Yamada, Masaki Hata, Hiroshi Nishiura, Yoshimitsu Abiko, Nobuyuki Terada, Keiji Nakasho
ABSTRACT
Interleukin 12 receptor β chain (IL12RB2) is a crucial regulatory factor involved in cell-mediated immune responses, and genetic variants of the gene encoding IL12RB2 are associated with susceptibility to various immune-related diseases. We previously demonstrated that haplotypes with single nucleotide polymorphisms (SNPs) in the 5' flanking region of IL12RB2, including -1035A>G (rs3762315) and -1023A>G (rs3762316), affect the expression of IL12RB2, thereby altering susceptibility to leprosy and periodontal diseases. In the present study, we identified transcription factors associated with the haplotype-specific transcriptional activity of IL12RB2 in T cells and NK cells. The -1023G polymorphism was found to create a consensus binding site for the transcription factor activating protein (AP)-1, and enzyme-linked immunosorbent assay (ELISA)-based binding assays showed that these SNPs enhanced AP-1 binding to this region. In reporter assays, suppression of JunB expression using siRNA eliminated differences in the -1035G/-1023G and -1035A/-1023A regions containing IL12RB2 promoter activity in Jurkat T cells and NK3.3 cells. These results suggested that the -1035/-1023 polymorphisms created differential binding affinities for JunB that could lead to differential IL12RB2 expression. Moreover, the -1035G and -1035A alleles formed binding sites for GATA-3 and myocyte enhancer factor-2 (MEF-2), respectively. Our data indicated that in addition to JunB, the SNP at -1035/-1023 influenced GATA-3 and MEF-2 binding affinity, potentially altering IL12RB2 transcriptional activity. These findings confirm the effects of rs3762315 and rs3762316 on IL12RB2 transcription. These genetic variants may alter cellular activation of T cells and NK cells and modify cell-mediated immune responses.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Human IL-12 R beta 2 ELISA, for serum, plasma and cell culture supernatants