Merck
  • Bioisosteric replacement of the alpha-amino carboxylic acid functionality in 2-amino-5-phosphonopentanoic acid yields unique 3,4-diamino-3-cyclobutene-1,2-dione containing NMDA antagonists.
Bioisosteric replacement of the alpha-amino carboxylic acid functionality in 2-amino-5-phosphonopentanoic acid yields unique 3,4-diamino-3-cyclobutene-1,2-dione containing NMDA antagonists.
Journal of medicinal chemistry (1992-12-21)
W A Kinney, N E Lee, D T Garrison, E J Podlesny, J T Simmonds, D Bramlett, R R Notvest, D M Kowal, R P Tasse
ABSTRACT
In this report, a novel bioisostere of the alpha-amino acid, 3,4-diamino-3-cyclobutene-1,2-dione, has been incorporated into a series of compounds which are NMDA antagonists. These compounds, which are achiral and easily prepared, demonstrated good affinity at the NMDA receptor by their ability to displace [3H]CPP binding in vitro. In particular, the phosphonic acid 24 provided protection against NMDA-induced lethality in mice equivalent to 2-amino-7-phosphonoheptanoic acid (5). This was considered an encouraging result in lieu of the fact that 24, like 5, lacks the conformational rigidity of the more potent NMDA antagonists. In addition, analogs that incorporate the 1,2,4-oxadiazolidine-3,5-dione heterocycle of quisqualic acid and the unsaturation of kainic acid were prepared to explore selectivity at the non-NMDA receptor subtypes.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
3,4-Diethoxy-3-cyclobutene-1,2-dione, 98%
Prezzi e disponibilità al momento non sono disponibili