Merck
[Experimental cryptorchism].
Ceskoslovenska pediatrie (1990-09-01)
J Vrabec, K Cervencík
ABSTRACT
The authors evaluate the effect of hormonal treatment with chorionic gonadotropin on histological structures of the testes in experimental work and in artificially induced cryptorchism in the rabbit; they evaluate histological findings in cryptorchid and normally descended testes.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Rhenium(V) chloride