Merck
Cisplatin resistance by induction of aldo-keto reductase family 1 member C2 in human bladder cancer cells.
Oncology letters (2014-02-15)
Akitomi Shirato, Tadahiko Kikugawa, Noriyoshi Miura, Nozomu Tanji, Nobuaki Takemori, Shigeki Higashiyama, Masayoshi Yokoyama
ABSTRACT
Cisplatin is currently the most effective anti-tumor agent available against bladder cancer. To clarify the mechanism underlying cisplatin resistance in bladder cancer, the present study examined the role of the aldo-keto reductase family 1 member C2 (AKR1C2) protein on chemoresistance using a human bladder cancer cell line. The function of AKR1C2 in chemoresistance was studied using the human HT1376 bladder cancer cell line and the cisplatin-resistant HT1376-CisR subline. AKR1C2 was expressed in HT1376-CisR cells, but not in the parental cells. The effect of small interfering (si) RNAs and an inhibitor targeting AKR1C2 was examined to determine whether cisplatin sensitivity can be rescued by blocking AKR1C2 expression or function. Silencing of AKR1C2 mRNA or inhibition of AKR1C2 by 5β-cholanic acid resulted in a decrease in the survival of cells following cisplatin exposure. Intracellular accumulation of reactive oxygen species (ROS) was determined using a 2,7-dichlorodihydrofluorescein diacetate (H
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human AKR1C2