Merck
Endometriosis: treatment strategies.
Annals of the New York Academy of Sciences (2003-12-03)
Rafael F Valle, John J Sciarra
ABSTRACT
Endometriosis is often a perplexing medical condition for both the physician and the patient. Accordingly, development of treatment strategies based on the needs of the individual patient is highly desirable. Although endometriosis has been part of the clinical practice for almost a century, many questions remain relating to the relationship between endometriosis and infertility as well as endometriosis and pelvic pain. Endometriosis is a disease of reproductive-age women, and it is now well recognized that a genetic susceptibility appears probable. The prevalence in the general population has never been clearly established. Factors to consider in management include the age and reproductive desires of the patient, the stage of the disease, and, most importantly, the symptoms. Therapeutic options include no treatment, medical therapy, surgery, or combination therapy. Oral contraceptives, androgenic agents, progestins, and gonadotropin releasing hormone (GnRH) analogs have all been used successfully, although at the present time, the latter preparations are the most popular medical therapy for endometriosis. Leuprolide acetate, goserelin acetate, and nafarelin acetate are all effective agents. Surgical therapy is appropriate, especially for advanced stages of the disease. Laparoscopy is an effective surgical approach with the goal of excision of visible endometriosis in a hemostatic fashion. Since endometriosis is a chronic condition, it is not uncommon for recurrences to occur. While endometriosis remains an enigmatic disease, the introduction of new pharmacologic agents, such as GnRH analogs and newer endoscopic methods of surgical treatment, have facilitated and improved the overall management of this disease.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Nafarelin acetate salt hydrate, ≥95% (HPLC), solid