Merck
Gd(III)-DOTA-modified sonosensitive liposomes for ultrasound-triggered release and MR imaging.
Nanoscale research letters (2012-08-21)
Suk Hyun Jung, Kyunga Na, Seul A Lee, Sun Hang Cho, Hasoo Seong, Byung Cheol Shin
ABSTRACT
Ultrasound-sensitive (sonosensitive) liposomes for tumor targeting have been studied in order to increase the antitumor efficacy of drugs and decrease the associated severe side effects. Liposomal contrast agents having Gd(III) are known as a nano-contrast agent system for the efficient and selective delivery of contrast agents into pathological sites. The objective of this study was to prepare Gd(III)-DOTA-modified sonosensitive liposomes (GdSL), which could deliver a model drug, doxorubicin (DOX), to a specific site and, at the same time, be capable of magnetic resonance (MR) imaging. The GdSL was prepared using synthesized Gd(III)-DOTA-1,2-distearoyl-sn-glycero-3-phosphoethanolamine lipid. Sonosensitivity of GdSL to 20-kHz ultrasound induced 33% to 40% of DOX release. The relaxivities (r1) of GdSL were 6.6 to 7.8 mM-1 s-1, which were higher than that of MR-bester®. Intracellular uptake properties of GdSL were evaluated according to the intensity of ultrasound. Intracellular uptake of DOX for ultrasound-triggered GdSL was higher than that for non-ultrasound-triggered GdSL. The results of our study suggest that the paramagnetic and sonosensitive liposomes, GdSL, may provide a versatile platform for molecular imaging and targeted drug delivery.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
1,2-Distearoyl-sn-glycero-3-phosphoethanolamine, ≥99%
Sigma-Aldrich
N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)uronium hexafluorophosphate, ≥99.0% (TLC/N)