Merck
  • Matrix softness regulates plasticity of tumour-repopulating cells via H3K9 demethylation and Sox2 expression.
Matrix softness regulates plasticity of tumour-repopulating cells via H3K9 demethylation and Sox2 expression.
Nature communications (2014-08-08)
Youhua Tan, Arash Tajik, Junwei Chen, Qiong Jia, Farhan Chowdhury, Lili Wang, Junjian Chen, Shuang Zhang, Ying Hong, Haiying Yi, Douglas C Wu, Yuejin Zhang, Fuxiang Wei, Yeh-Chuin Poh, Jihye Seong, Rishi Singh, Li-Jung Lin, Sultan Doğanay, Yong Li, Haibo Jia, Taekjip Ha, Yingxiao Wang, Bo Huang, Ning Wang
ABSTRACT
Tumour-repopulating cells (TRCs) are a self-renewing, tumorigenic subpopulation of cancer cells critical in cancer progression. However, the underlying mechanisms of how TRCs maintain their self-renewing capability remain elusive. Here we show that relatively undifferentiated melanoma TRCs exhibit plasticity in Cdc42-mediated mechanical stiffening, histone 3 lysine residue 9 (H3K9) methylation, Sox2 expression and self-renewal capability. In contrast to differentiated melanoma cells, TRCs have a low level of H3K9 methylation that is unresponsive to matrix stiffness or applied forces. Silencing H3K9 methyltransferase G9a or SUV39h1 elevates the self-renewal capability of differentiated melanoma cells in a Sox2-dependent manner. Mechanistically, H3K9 methylation at the Sox2 promoter region inhibits Sox2 expression that is essential in maintaining self-renewal and tumorigenicity of TRCs both in vitro and in vivo. Taken together, our data suggest that 3D soft-fibrin-matrix-mediated cell softening, H3K9 demethylation and Sox2 gene expression are essential in regulating TRC self-renewal.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
ChIPAb+ Trimethyl-Histone H3 (Lys9) - ChIP Validated Antibody and Primer Set, from rabbit
Sigma-Aldrich
ChIPAb+ Dimethyl-Histone H3 (Lys9) - ChIP Validated Antibody and Primer Set, serum, from rabbit