Merck
Nickel-catalyzed thioallylation of alkynes with allyl phenyl sulfides.
Organic letters (2007-01-16)
Ruimao Hua, Hideaki Takeda, Shun-ya Onozawa, Yoshimoto Abe, Masato Tanaka
ABSTRACT
Allylic sulfides add to alkynes in the presence of nickel complexes efficiently to afford thio-1,4-dienes regio- and stereoselectively. Functional groups such as alkoxy, siloxy, hydroxy, carboalkoxy, chloro, and cyano groups are tolerated. A mechanism that involves a pi-allyl nickel intermediate is proposed on the basis of isolation of pi-allyl complexes and distribution of products in the reactions of alpha- or gamma-methylated allyl sulfide. [reaction: see text].
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Bis(1,5-cyclooctadiene)nickel(0)