Merck
Pt-free carbon-based fuel cell catalyst prepared from spherical polyimide for enhanced oxygen diffusion.
Scientific reports (2016-03-19)
Yuta Nabae, Shinsuke Nagata, Teruaki Hayakawa, Hideharu Niwa, Yoshihisa Harada, Masaharu Oshima, Ayano Isoda, Atsushi Matsunaga, Kazuhisa Tanaka, Tsutomu Aoki
ABSTRACT
The development of a non-precious metal (NPM) fuel cell catalyst is extremely important to achieve globalization of polymer electrolyte fuel cells due to the cost and scarcity of platinum. Here, we report on a NPM cathode catalyst prepared by the pyrolysis of spherical polyimide nanoparticles that contain small amounts of Fe additive. 60 nm diameter Fe-containing polyimide nanoparticles were successfully synthesized by the precipitation polymerization of pyromellitic acid dianhydride and 1,3,5-tris(4-aminophenyl)benzene with Fe(acac)3 (acac = acetylacetonate) as an additive. The particles were subsequently carbonized by multistep pyrolysis to obtain the NPM catalyst while retaining the small particle size. The catalyst has good performance and promising durability for fuel cell applications. The fuel cell performance under a 0.2 MPa air atmosphere at 80 °C of 1.0 A cm(-2) at 0.46 V is especially remarkable and better than that previously reported.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Pyromellitic dianhydride, 97%