Merck
PolyMetformin combines carrier and anticancer activities for in vivo siRNA delivery.
Nature communications (2016-06-07)
Yi Zhao, Wei Wang, Shutao Guo, Yuhua Wang, Lei Miao, Yang Xiong, Leaf Huang
ABSTRACT
Metformin, a widely implemented anti-diabetic drug, exhibits potent anticancer efficacies. Herein a polymeric construction of Metformin, PolyMetformin (PolyMet) is successfully synthesized through conjugation of linear polyethylenimine (PEI) with dicyandiamide. The delocalization of cationic charges in the biguanide groups of PolyMet reduces the toxicity of PEI both in vitro and in vivo. Furthermore, the polycationic properties of PolyMet permits capture of siRNA into a core-membrane structured lipid-polycation-hyaluronic acid (LPH) nanoparticle for systemic gene delivery. Advances herein permit LPH-PolyMet nanoparticles to facilitate VEGF siRNA delivery for VEGF knockdown in a human lung cancer xenograft, leading to enhanced tumour suppressive efficacy. Even in the absence of RNAi, LPH-PolyMet nanoparticles act similarly to Metformin and induce antitumour efficacy through activation of the AMPK and inhibition of the mTOR. In essence, PolyMet successfully combines the intrinsic anticancer efficacy of Metformin with the capacity to carry siRNA to enhance the therapeutic activity of an anticancer gene therapy.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Avanti
18:1 TAP (DOTAP), Avanti Research - A Croda Brand
Avanti
DSPE-PEG(2000) Amine, Avanti Research - A Croda Brand
Avanti
18:1 TAP (DOTAP), Avanti Research - A Croda Brand
Avanti
DSPE-PEG(2000) Amine, Avanti Research - A Croda Brand 880128C