- Upregulation of long non-coding RNA LINC00152 by SP1 contributes to gallbladder cancer cell growth and tumor metastasis via PI3K/AKT pathway.
Upregulation of long non-coding RNA LINC00152 by SP1 contributes to gallbladder cancer cell growth and tumor metastasis via PI3K/AKT pathway.
American journal of translational research (2016-11-11)
Qiang Cai, Zhen-Qiang Wang, Shou-Hua Wang, Chen Li, Zheng-Gang Zhu, Zhi-Wei Quan, Wen-Jie Zhang
PMID27829993
ABSTRACT
Gallbladder cancer (GBC) is one of the most lethal cancers with poor prognosis. In this study, we report that the long non-coding RNA LINC00152 is significantly upregulated in GBC tissues and cell lines. The high LINC00152 levels correlated positively with tumor status progression, lymph node invasion and TNM stage advancement. Functionally, we revealed that LINC00152 dramatically promoted cell proliferation, metastasis and inhibited apoptosis
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto