Merck
  • Monosialotetrahexosylganglioside protect cerebral ischemia/reperfusion injury through upregulating the expression of tyrosine hydroxylase by inhibiting lipid peroxidation.
Monosialotetrahexosylganglioside protect cerebral ischemia/reperfusion injury through upregulating the expression of tyrosine hydroxylase by inhibiting lipid peroxidation.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2016-11-17)
Li-Mao, Yin-Juan Liao, Guang-Han Hou, Zhong-Bao Yang, Mei-Ling Zuo
ABSTRACT
To explore the new mechanism of neuroprtection of monosialotetrahexosylganglioside and providing reliable theoretical foundation and experimental evidence for the emergency treatment and rehabilitation of cerebral ischemia/reperfusion injury. A rat model of cerebral ischemia/reperfusion injury was constructed and intervened with monosialotetrahexosylganglioside(5mg/kg) and lipid peroxidation inhibitor U-101033E(40mg/kg). TTC straining and neurobiological function score were used to evaluate brain injury. 4-HNE and MDA content were measured to evaluate lipid peroxidation. The expression of tyrosine hydroxilase at both mRNA and protein levels and enzyme activity were determined to evaluate the gene disfunction. Tyrosine content in brain and in serum and the DOPA content in plasma were measured to evaluate the metabolism of tyrosine. As the study shown, cerebral ischemia/reperfusion lead to brain infarction and neurobiological function losing accompany with upregulation of 4-HNE and MDA levels and downregulation of TH expression (mRNA and protein) and decreased enzyme activity. The results above mentioned can be reversed obviously by intervening with monosialotetrahexosylganglioside and lipid peroxidation inhibitor U-101033E. Toxic aldehyde accumulation leaded to disfunction of tyrosine hydroxylase and excessive tyrosine and decreased synthesis of catecholamine neurotransmitter such as dopamine and accelerated neuron cell injury. Both monosialotetrahexosylganglioside and U-101033E presented neuroprotecion by restoring the tyrosine/dopa pathway through reversing the function of tyrosine hydroxylase by inhibiting lipid peroxidation.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Tyrosine Colorimetric Assay Kit, Sufficient for 100 Colorimetric tests