Merck
p53R2 regulates thioredoxin reductase activity through interaction with TrxR2.
Biochemical and biophysical research communications (2016-11-22)
Seon-Joo Park, Hong Beum Kim, Chunmei Piao, Mi Yeong Kang, Sang-Gon Park, Seok Won Kim, Jung-Hee Lee
ABSTRACT
Ribonucleotide reductase small subunit p53R2 is a member of the ribonucleotide reductase family that supplies dNTPs for nuclear and mitochondrial DNA replication and repair. Here, we have identified a mitochondrial thioredoxin reductase 2 (TrxR2) as a novel p53R2-binding protein. We demonstrated a direct interaction between the two, and observed that p53R2 stimulated the enzymatic activity of TrxR in vitro. Moreover, TrxR2 activity was significantly lower in p53R2 knockdown cells, and increased when p53R2 was overexpressed, effects that were independent of p53. Furthermore, p53R2 knockdown suppressed UV-induced TrxR activity. These findings suggest that p53R2 acts as a positive regulator of TrxR2 activity in mitochondria both under normal physiological conditions and during the cellular response to DNA damage.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human RRM2B