Merck
Routing of the RAB6 secretory pathway towards the lysosome related organelle of melanocytes.
Nature communications (2017-06-14)
Anand Patwardhan, Sabine Bardin, Stéphanie Miserey-Lenkei, Lionel Larue, Bruno Goud, Graça Raposo, Cédric Delevoye
ABSTRACT
Exocytic carriers convey neo-synthesized components from the Golgi apparatus to the cell surface. While the release and anterograde movement of Golgi-derived vesicles require the small GTPase RAB6, its effector ELKS promotes the targeting and docking of secretory vesicles to particular areas of the plasma membrane. Here, we show that specialized cell types exploit and divert the secretory pathway towards lysosome related organelles. In cultured melanocytes, the secretory route relies on RAB6 and ELKS to directly transport and dock Golgi-derived carriers to melanosomes. By delivering specific cargos, such as MART-1 and TYRP2/ DCT, the RAB6/ELKS-dependent secretory pathway controls the formation and maturation of melanosomes but also pigment synthesis. In addition, pigmentation defects are observed in RAB6 KO mice. Our data together reveal for the first time that the secretory pathway can be directed towards intracellular organelles of endosomal origin to ensure their biogenesis and function.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-actinaβ monoclonale, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-GAPDH, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting human RAB6A