Merck
Vapor-phase-synthesized fluoroacrylate polymer thin films: thermal stability and structural properties.
Beilstein journal of nanotechnology (2017-05-27)
Paul Christian, Anna Maria Coclite
ABSTRACT
In this study, the thermal, chemical and structural stability of 1
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Etilenglicole dimetilacrilato, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Luperox® DI, tert-Butyl peroxide, 98%
Sigma-Aldrich
1H,1H,2H,2H-Perfluorodecyl acrylate, contains 100 ppm tert-butylcatechol as inhibitor, 97%