Merck
Brain Tissue Reaction to Deep Brain Stimulation-A Longitudinal Study of DBS in the Goettingen Minipig.
Neuromodulation : journal of the International Neuromodulation Society (2017-02-22)
Dariusz Orlowski, Alexandre Michalis, Andreas N Glud, Anders R Korshøj, Lise M Fitting, Trine W Mikkelsen, André Mercanzini, Alain Jordan, Alain Dransart, Jens C H Sørensen
ABSTRACT
The use of Deep Brain Stimulation (DBS) in treatment of various brain disorders is constantly growing; however, the number of studies of the reaction of the brain tissue toward implanted leads is still limited. Therefore, the aim of our study was to analyze the impact of DBS leads on brain tissue in a large animal model using minipigs. Twelve female animals, one control and eleven with bilaterally implanted DBS electrodes were used in our experiment. 3, 6, and 12 months after implantation the animals were sacrificed, perfused and the brains were removed. Tissue blocks containing the lead tracks were dissected, frozen, sectioned into 40 µm sections and stained using Nissl and Eosin, anti-GFAPab or Isolectin. The tissue reaction was analyzed at five levels, following from the distal lead tip, to compare tissue response in stimulated and nonstimulated areas: four segments along each level of electrodes, and the fifth level lying outside the electrode area (control area). The sections were described both qualitatively and quantitatively. Quantitative assessment of the reaction to the implanted electrode was based on the measurement of the area covered by the staining and the thickness of the glial scar. Tissue reaction was, on average, limited to distance of 500 μm from the lead track. The tissue response after 12 months was weaker than after 6 months confirming that it stabilizes over a time. There was no histological evidence that the stimulated part of the electrode triggered different tissue response than its nonstimulated part.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia), Isolectin B4 (BSI-B4), peroxidase conjugate, lyophilized powder