Merck
Whole-Genome and Epigenomic Landscapes of Etiologically Distinct Subtypes of Cholangiocarcinoma.
Cancer discovery (2017-07-02)
Apinya Jusakul, Ioana Cutcutache, Chern Han Yong, Jing Quan Lim, Mi Ni Huang, Nisha Padmanabhan, Vishwa Nellore, Sarinya Kongpetch, Alvin Wei Tian Ng, Ley Moy Ng, Su Pin Choo, Swe Swe Myint, Raynoo Thanan, Sanjanaa Nagarajan, Weng Khong Lim, Cedric Chuan Young Ng, Arnoud Boot, Mo Liu, Choon Kiat Ong, Vikneswari Rajasegaran, Stefanus Lie, Alvin Soon Tiong Lim, Tse Hui Lim, Jing Tan, Jia Liang Loh, John R McPherson, Narong Khuntikeo, Vajaraphongsa Bhudhisawasdi, Puangrat Yongvanit, Sopit Wongkham, Yasushi Totoki, Hiromi Nakamura, Yasuhito Arai, Satoshi Yamasaki, Pierce Kah-Hoe Chow, Alexander Yaw Fui Chung, London Lucien Peng Jin Ooi, Kiat Hon Lim, Simona Dima, Dan G Duda, Irinel Popescu, Philippe Broet, Sen-Yung Hsieh, Ming-Chin Yu, Aldo Scarpa, Jiaming Lai, Di-Xian Luo, André Lopes Carvalho, André Luiz Vettore, Hyungjin Rhee, Young Nyun Park, Ludmil B Alexandrov, Raluca Gordân, Steven G Rozen, Tatsuhiro Shibata, Chawalit Pairojkul, Bin Tean Teh, Patrick Tan
ABSTRACT
Cholangiocarcinoma (CCA) is a hepatobiliary malignancy exhibiting high incidence in countries with endemic liver-fluke infection. We analyzed 489 CCAs from 10 countries, combining whole-genome (71 cases), targeted/exome, copy-number, gene expression, and DNA methylation information. Integrative clustering defined 4 CCA clusters-fluke-positive CCAs (clusters 1/2) are enriched in
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human RASA1