Merck
Accessing Stable Magnesium Acyl Compounds: Reductive Cleavage of Esters by Magnesium(I) Dimers.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-09-07)
Aaron J Boutland, Carlos A Lamsfus, Brant Maitland, Laurent Maron, Andreas Stasch, Cameron Jones
ABSTRACT
The first examples of magnesium acyls, [(Nacnac)Mg{μ-C(Ph)O}(μ-OR)Mg(Nacnac)] (R=Me, tBu or Ph; Nacnac=[HC(MeCNAr)
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Nitrogen-14N2, 99.99 atom % 14N