Merck
Lateral flow assay for carbohydrate antigen 19-9 in whole blood by using magnetized carbon nanotubes.
Mikrochimica acta (2017-12-05)
Yan Huang, Yongqiang Wen, Kwaku Baryeh, Sunitha Takalkar, Michelle Lund, Xueji Zhang, Guodong Liu
ABSTRACT
The authors describe a rapid, low-cost and sensitive approach for the determination of carbohydrate antigen 19-9 (CA 19-9) in whole blood by using magnetized carbon nanotube (MCNT) and lateral flow strip biosensor (LFSB). MCNTs were synthesized by depositing magnetite (Fe
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
N-idrossisolfosuccinimide, ≥98% (HPLC)
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)