Merck
The SH2B3 and KCNK5 loci may be implicated in regulation of platelet count, volume, and maturity.
Thrombosis research (2017-09-03)
Morten K Christiansen, Sanne B Larsen, Mette Nyegaard, Søs Neergaard-Petersen, Morten Würtz, Erik L Grove, Anne-Mette Hvas, Henrik K Jensen, Steen D Kristensen
ABSTRACT
In recent genome-wide association studies, coronary artery disease (CAD) and myocardial infarction (MI) have been linked to a number of genetic variants, but their role in thrombopoiesis is largely unknown. We investigated the association between CAD and MI-associated genetic variants and five thrombopoiesis-related indices: platelet count (PC), mean platelet volume (MPV), immature platelet count (IPC), immature platelet fraction (IPF), and serum thrombopoietin (TPO). We genotyped 45 genome-wide significant CAD/MI-markers in 879 stable CAD patients. A genetic risk score was calculated to assess the combined risk associated with all the genetic variants. Platelet indices were analysed using the Sysmex XE-2100 haematology analyser. TPO was measured by ELISA. Two variants were nominally associated with several indices; for rs10947789 (KCNK5), the adjusted geometric mean was 2% higher for MPV (95% confidence interval: 1-2%, p=0.002), 6% for IPC (0-12%, p=0.033), and 9% for IPF (3-16%, p=0.004) per CAD risk allele. Moreover, an 11% lower TPO (3-19%, p=0.010) was observed. Rs3184504 (SH2B3) was associated with a higher adjusted geometric mean of 3% (1-6%, p=0.003) per CAD risk allele for PC, and an 11% (5-17%, p<0.001) lower TPO. Furthermore, the adjusted IPC was 5% (0-9%, p=0.037) lower per CAD risk allele for PC, whereas IPF levels did not vary across genotypes. As a novel finding, our study suggests a role for KCNK5 in the regulation of platelet size and maturity. Furthermore, our findings confirm an association between the SH2B3-locus and platelet count.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Human TPO / Thyroid Peroxidase ELISA Kit, for serum, plasma, cell culture supernatants and urine