Merck
The Electro-Deposition/Dissolution of CuSO
The journal of physical chemistry. B (2017-10-19)
Juan-Jesús Velasco-Vélez, Katarzyna Skorupska, Elias Frei, Yu-Cheng Huang, Chung-Li Dong, Bing-Jian Su, Cheng-Jhih Hsu, Hung-Yu Chou, Jin-Ming Chen, Peter Strasser, Robert Schlögl, Axel Knop-Gericke, Cheng-Hao Chuang
ABSTRACT
The electrodeposition nature of copper on a gold electrode in a 4.8 pH CuSO
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
N,N′-Bis(3-methylphenyl)-N,N′-diphenylbenzidine, 99%