Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
1 L | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 1 590,00 Kč |
2 L | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 2 720,00 Kč |
4 x 2 L | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 8 170,00 Kč |
6 x 1 L | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 9 080,00 Kč |
12 x 100 mL | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 10 200,00 Kč |
O této položce
1 590,00 Kč
Quality Segment
grade
anhydrous
vapor density
1.1 (vs air), 1.11 (vs air)
vapor pressure
410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)
description
Various Returnable Container Options
product line
ISOM8™
assay
≥99.75%
form
liquid
autoignition temp.
433 °C (811°F), 725 °F
shelf life
60 mo.
expl. lim.
36 %
packaging
Sure/Seal™ of 1 L, Sure/Seal™ of 6 × 1 L, Sure/Seal™ of 12 × 100 mL, Sure/Seal™ of 4 × 2 L
technique(s)
tissue processing: suitable
impurities
<0.005% water
evapn. residue
<0.0005%
refractive index
n/D 1.3290 (lit.), n20/D 1.329 (lit.)
pH
7 (20 °C, 10 g/L)
bp
64.7 °C
mp
-98 °C, −98 °C (lit.)
transition temp
flash point 13 °C (55°F)
solubility
miscible
density
0.791 g/mL at 20 °C, 0.791 g/mL at 25 °C (lit.)
cation traces
(See Dossier)
suitability
suitable for (histological solvents), suitable for fixing solution
application(s)
diagnostic assay manufacturing
life science and biopharma
SMILES string
CO
InChI
1S/CH4O/c1-2/h2H,1H3
InChI key
OKKJLVBELUTLKV-UHFFFAOYSA-N
General description
Our portfolio of ISOM8 products and chemistries is ever evolving to support our valued Sigma-Aldrich® customers′ requirements. Please navigate to our documents section to look at dossiers and other quality documentation for this product, provided with support of our Elevated Assurance Program.
Our ISOM8™ Methanol is available within the USA with our ReCycler® Solvent System for returnable, reusable, and readily available bulk solvent delivery, to reduce packaging waste and help you achieve your sustainablility goals. To request ISOM8 and other product lines in returnable containers Click Here.
Application
Methanol is widely used in cell fixation processes, preserving cellular structures and morphology for microscopy, immunohistochemistry, and other analytical techniques. Its ability to precipitate proteins and denature nucleic acids makes it an essential tool in biological and biomedical research.
Chemical Synthesis
As a versatile building block in organic synthesis, non-GMP anhydrous methanol serves as a solvent and reagent in the production of pharmaceuticals, agrochemicals, and specialty chemicals, facilitating efficient and effective reactions.
Formulation Development
In the formulation of coatings, adhesives, and sealants, non-GMP anhydrous methanol acts as a solvent, improving the application properties and performance of end products.
Features and Benefits
- High Purity: Our anhydrous methanol is produced to meet stringent quality standards, ensuring minimal impurities for reliable performance in sensitive applications.
- Effective Fixative: Serves as a reliable fixative for biological samples, preserving cellular structures for microscopy and analytical techniques.
- Versatile Applications: Suitable for a wide range of applications including synthesis, extraction, and formulation development.
- Returnable Containers: Offered in returnable containers, promoting sustainability and responsible resource management.
- Reliable Results: The high purity and low water content ensure consistent and reproducible results in versatile applications.
- Broad Applicability: Suitable for various industries including pharmaceuticals, biotechnology, energy, and manufacturing, making it a valuable addition to your supply chain.
- Cost Efficiency: Returnable containers can lead to cost savings over time, as customers can reuse them rather than purchasing new packaging for each order.
Packaging
For bulk amounts in US, we offer returnables through ReCycler® Program.
Legal Information
1 of 1
Tato položka | ||
|---|---|---|
| description ≥99.75%, ISOM8™, anhydrous | description Absolute - Acetone free | description ACS reagent, ≥99.8% |
| technique(s) tissue processing: suitable | technique(s) - | technique(s) - |
| application(s) diagnostic assay manufacturing | application(s) - | application(s) - |
| Quality Level 300 | Quality Level 200 | Quality Level 200, 100 |
| grade anhydrous | grade - | grade ACS reagent |
| assay ≥99.75% | assay - | assay ≥99.8% |
| solubility miscible | solubility - | solubility - |
Still not finding the right product?
Explore all of our products under Methanol
Výstražné slovo
Danger
Kódy nebezpečí
Bezpečnostní pokyny
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Cílové orgány
Eyes,Central nervous system
Skladovací třída
3 - Flammable liquids
Bod vzplanutí (°F)
49.5 °F - closed cup
Bod vzplanutí (°C)
9.7 °C - closed cup
Seznamy omezení vyplývajících z předpisů
Seznamy omezení vyplývajících z předpisů jsou poskytovány zejména pro chemické produkty. Pro jiné než chemické produkty lze poskytovat pouze omezené informace. Žádný záznam znamená, že v seznamech není obsažena žádná ze součástí. Povinností uživatele je zajistit bezpečné a zákonné použití produktu.
67-56-1
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?
Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.



