Přejít k obsahu
Merck
Docking study yields four novel inhibitors of the protooncogene Pim-1 kinase.
Journal of medicinal chemistry (2008-02-23)
Albert C Pierce, Marc Jacobs, Cameron Stuver-Moody
ANOTACE
To supplement the hits from a high throughput screen, docking was performed against Pim-1 kinase. Glide docking was augmented with a filter to require traditional or aromatic CH..O hydrogen bonds to the kinase hinge. Four diverse actives, of 96 molecules assayed, had K(i) values between 0.091 and 4.5 microM. This gives a 14-fold enrichment over the earlier HTS run, and the two crystal structures solved confirmed the binding modes predicted by docking.