Přejít k obsahu
Merck
  • Inhibition of protein tyrosine phosphatase-1B with antisense oligonucleotides improves insulin sensitivity and increases adiponectin concentrations in monkeys.
Inhibition of protein tyrosine phosphatase-1B with antisense oligonucleotides improves insulin sensitivity and increases adiponectin concentrations in monkeys.
Endocrinology (2009-01-24)
Michael M Swarbrick, Peter J Havel, Arthur A Levin, Andrew A Bremer, Kimber L Stanhope, Madeline Butler, Sheri L Booten, James L Graham, Robert A McKay, Susan F Murray, Lynnetta M Watts, Brett P Monia, Sanjay Bhanot
ANOTACE
Protein tyrosine phosphatase (PTP)-1B antagonizes insulin signaling and is a potential therapeutic target for insulin resistance associated with obesity and type 2 diabetes. To date, studies of PTP-1B have been limited by the availability of specific antagonists; however, treatment of rodents with antisense oligonucleotides (ASOs) directed against PTP-1B improves insulin sensitivity, inhibits lipogenic gene expression, and reduces triglyceride accumulation in liver and adipose tissue. Here we investigated ASO-mediated PTP-1B inhibition in primates. First, PTP-1B ASO (ISIS 113715) dose-dependently inhibited PTP-1B mRNA and protein expression in cultured monkey hepatocytes. Subcutaneous administration of ISIS 113715 reduced PTP-1B mRNA expression in liver and adipose tissue of normal-weight monkeys by 40-50% and improved insulin sensitivity during an iv glucose tolerance test (IVGTT). In obese, insulin-resistant rhesus monkeys, treatment with 20 mg/kg ISIS 113715 for 4 wk reduced fasting concentrations of insulin and glucose and reduced insulin responses during an IVGTT. In these animals, adiponectin concentrations were also increased by 70%, most of which was an increase of high-molecular-weight oligomers. These effects were not observed in monkeys on a lower, dose-escalation regimen (1-10 mg/kg over 9 wk). Overall, the increase of adiponectin concentrations during ISIS 113715 treatment was correlated with the lowering of insulin responses during IVGTT (r = -0.47, P = 0.042). These results indicate that inhibition of PTP-1B with ASOs such as ISIS 113715 may be a viable approach for the treatment and prevention of obesity-associated insulin resistance and type 2 diabetes because they potently increase adiponectin concentrations in addition to improving insulin sensitivity.