- An efficient computational model to predict the synthetic utility of heterocyclic arynes.
An efficient computational model to predict the synthetic utility of heterocyclic arynes.
Angewandte Chemie (International ed. in English) (2012-02-07)
Adam E Goetz, Sarah M Bronner, Jordan D Cisneros, Joshua M Melamed, Robert S Paton, K N Houk, Neil K Garg
PMID22307724
ANOTACE
Think before you act: a computational approach is reported for evaluating the synthetic potential of heterocyclic arynes. Routine and rapid calculations of arene dehydrogenation energies and aryne angle distortion predict the likelihood that a given hetaryne can be generated, as well as the degree of regioselectivity expected in a reaction between a given hetaryne and a nucleophilic trapping agent.