Merck
Prenatal exposure to bisphenol A disrupts mouse fetal lung development.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-08-19)
Ayten Hijazi, Haiyan Guan, Maria Cernea, Kaiping Yang
ABSTRACT
Developmental exposure to bisphenol A (BPA) is associated with lung dysfunction and diseases. However, it is unknown if this association has a fetal origin. The present study addressed this important question by examining the effects of BPA on fetal lung development. BPA was administered to pregnant mice via diet from embryonic day (E) 7.5 to E18.5. Fetal lungs were analyzed at E18.5 for changes in structure and expression of key molecular markers of lung maturation. Our main findings were as follows: BPA severely retards fetal lung maturation, as evidenced by diminished alveolar airspace (15% of control) and thickened septa, hallmarks of lung immaturity; this immaturity is characterized by aberrant alveolar epithelial type I cell differentiation because expression of the type I cell marker, aquaporin 5, but not type II cell markers, is dramatically reduced (16% of control); and the effects of BPA are likely mediated through the glucocorticoid signaling pathway because the expression of epithelial sodium channel γ and glutathione peroxidase, 2 well-known glucocorticoid target genes, is down-regulated in BPA-exposed fetal lungs, and, importantly, maternal dexamethasone administration rescues the lung immaturity phenotype. Taken together, these findings demonstrate that BPA disrupts fetal lung maturation, thus suggesting a fetal origin for BPA-induced lung diseases.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder