Merck
Novel hydroxyapatite biomaterial covalently linked to raloxifene.
International journal of immunopathology and pharmacology (2014-10-04)
L Meme, A Santarelli, G Marzo, M Emanuelli, P F Nocini, D Bertossi, A Putignano, M Dioguardi, L Lo Muzio, F Bambini
RESUMEN
Since raloxifene, a drug used in osteoporosis therapy, inhibits osteoclast, but not osteoblast functions, it has been suggested to improve recovery during implant surgery. The present paper describes an effective method to link raloxifene, through a covalent bond, to a nano-Hydroxyapatite-based biomaterial by interfacing with (3-aminopropyl)-Triethoxysilane as assessed by Infra Red-Fourier Transformed (IR-FT) spectroscopy and Scanning Electron Microscope (SEM). To evaluate the safety of this modified new material, the vitality of osteoblast-like cells cultured with the new biomaterial was then investigated. Raloxifene-conjugated HAbiomaterial has been shown to be a safe material easy to obtain which could be an interesting starting point for the use of a new functional biomaterial suitable in bone regeneration procedures.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Alcohol isopropílico, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Alcohol isopropílico, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard