Merck
  • Quantitative study of proteomic alterations in a Zebrafish (danio rerio) cell line infected with the Singapore Grouper Iridovirus (SGIV).
Quantitative study of proteomic alterations in a Zebrafish (danio rerio) cell line infected with the Singapore Grouper Iridovirus (SGIV).
Virus research (2015-02-11)
Fan Wang, Yi Zhu, Choy Leong Hew
RESUMEN
We demonstrate, for the first time, that Singapore Grouper Iridovirus (SGIV) can successfully infect a Zebrafish cell line. Combined with the recent availability of the complete zebrafish (danio rerio) genome, this provides an opportunity to investigate virus-host interactions at the molecular level. Using iTRAQ labeling and two-dimensional LC/MS/MS quantitative proteomics, 157 zebrafish proteins exhibiting significant alterations in expression levels following SGIV infection were identified. Gene ontology analysis revealed that SGIV controls a wide aspect of zebrafish host machinery to ensure replication and propagation. In order to probe the mechanism underlying SGIV infection in Zebrafish cells, we used an anti-sense morpholino to knockdown orf86r, an immediate early viral gene that encodes the SGIV protein ORF86R. The expression profile of certain host proteins involved in replication was altered upon knockdown. In particular, expression of CNOT, a non-enzymatic subunit of the CCR4-NOT transcription complex was markedly affected. Taken together, these findings provide a new insight on the function of the essential viral protein ORF86R. Our results show that Singapore Grouper Iridovirus infection of a Zebrafish cell line is a useful new tool to study virus-host interactions.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido fórmico, reagent grade, ≥95%
Sigma-Aldrich
Ácido fórmico, ACS reagent, ≥96%
Sigma-Aldrich
2- Mercaptoetanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2- Mercaptoetanol, ≥99.0%
Sigma-Aldrich
L-Glutamina, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Ácido fórmico, ACS reagent, ≥88%
Sigma-Aldrich
L-Glutamina, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
2- Mercaptoetanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
SAFC
L-Glutamina
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamina, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ácido fórmico, ≥95%, FCC, FG
Sigma-Aldrich
L-Glutamina, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamina
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
Supelco
2- Mercaptoetanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Supelco
L-Glutamina, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
L-Glutamina, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)