Merck
  • A kinetic characterization of the gill V(H(+))-ATPase in juvenile and adult Macrobrachium amazonicum, a diadromous palaemonid shrimp.
A kinetic characterization of the gill V(H(+))-ATPase in juvenile and adult Macrobrachium amazonicum, a diadromous palaemonid shrimp.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology (2014-12-03)
Malson N Lucena, Marcelo R Pinto, Daniela P Garçon, John C McNamara, Francisco A Leone
RESUMEN
Novel kinetic properties of a microsomal gill V(H(+))-ATPase from juvenile and adult Amazon River shrimp, Macrobrachium amazonicum, are described. While protein expression patterns are markedly different, Western blot analysis reveals a sole immunoreactive band, suggesting a single V(H(+))-ATPase subunit isoform, distributed in membrane fractions of similar density in both ontogenetic stages. Immunofluorescence labeling locates the V(H(+))-ATPase in the apical regions of the lamellar pillar cells in both stages in which mRNA expression of the V(H(+))-ATPase B-subunit is identical. Juvenile (36.6±3.3 nmol Pi min(-1) mg(-1)) and adult (41.6±1.3 nmol Pi min(-1) mg(-1)) V(H(+))-ATPase activities are similar, the apparent affinity for ATP of the adult enzyme (K0.5=0.21±0.02 mmol L(-1)) being 3-fold greater than for juveniles (K0.5=0.61±0.01 mmol L(-1)). The K0.5 for Mg(2+) interaction with the juvenile V(H(+))-ATPase (1.40 ± 0.07 mmol L(-1)) is ≈6-fold greater than for adults (0.26±0.02 mmol L(-1)) while the bafilomycin A1 inhibition constant (KI) is 45.0±2.3 nmol L(-1) and 24.2±1.2 nmol L(-1), for juveniles and adults, respectively. Both stages exhibited residual bafilomycin-insensitive ATPase activity of ≈25 nmol Pi min(-1) mg(-1), suggesting the presence of ATPases other than the V(H(+))-ATPase. These differences may reflect a long-term regulatory mechanism of V(H(+))-ATPase activity, and suggest stage-specific enzyme modulation. This is the first kinetic analysis of V(H(+))-ATPase activity in different ontogenetic stages of a freshwater shrimp and allows better comprehension of the biochemical adaptations underpinning the establishment of palaemonid shrimps in fresh water.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Alcohol etílico puro 200, Molecular Biology
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Dimetilsulfóxido, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimetilsulfóxido, ACS reagent, ≥99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimetilsulfóxido, Molecular Biology
Sigma-Aldrich
Dimetilsulfóxido, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Alcohol etílico puro, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimetilsulfóxido, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimetilsulfóxido, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Alcohol etílico puro, 200 proof, meets USP testing specifications
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Dimetilsulfóxido, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Alcohol isopropílico, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Alcohol etílico puro 190, for molecular biology
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Trietanolamina, ≥99.0% (GC)
Sigma-Aldrich
Fluoruro de fenilmetansulfonilo, ≥98.5% (GC)
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Etanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Dimetilsulfóxido, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Alcohol isopropílico, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Etanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Fluoruro de fenilmetansulfonilo, ≥99.0% (T)
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)