Sign Into View Organizational & Contract Pricing
Select a Size
25 EA
Inquire
This item is not able to be purchased as a guest user. For further information, please contact us at +33 1 85 14 81 89or
.About This Item
UNSPSC Code:
40161507
eCl@ss:
32031602
Skip To
product line
Millipore Express®
feature
hydrophilic
manufacturer/tradename
Millipore Express®
filter diam.
47 mm
matrix
Millipore Express® SHF
pore size
0.2 μm nominal pore size
Legal Information
MILLIPORE EXPRESS is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service