Baris Sel untuk Bioproses
Baris sel yang mempertahankan karakteristik pertumbuhan dan produktivitas yang optimal sangat penting untuk proses manufaktur biopharmaceutical, baik Anda memproduksi antibodi monoklonal, protein rekombinan, atau vektor virus untuk terapi gen. Sistem dan platform baris sel siap pakai kami dapat mempercepat pengembangan hulu Anda dan secara signifikan mengurangi waktu, biaya, dan risiko dalam mengembangkan baris sel baru. Baris sel kami dirancang secara rasional dan sesuai dengan standar manufaktur bioteknologi.
Fitur utama meliputi:
- Baris sel yang sepenuhnya dapat dilacak dengan dokumentasi lengkap
- Disimpan dan dikarakterisasi sesuai cGMP
- Media dan nutrisi yang dipasangkan, dioptimalkan, dan didefinisikan secara kimia
- Protokol pengguna komprehensif untuk pengembangan garis sel yang efisien dan skalabilitas yang mulus, didukung oleh konsultasi teknis
Products
Sumber Daya Terkait
- Data Sheet: CHOZN® Elite Cell Line Product Information Sheet
CHOZN® Elite: : A production adapted host cell line to fulfill all your bioprocessing needs.
- Flyer: Sf-RVN® Platform
With the Sf-RVN® Platform, we provide a rhabdovirus-free cell line that improves the safety profile of your bioprocesses combined with a chemically defined medium specifically formulated to get excellent growth and productivity.
- Product Information Sheet: Sf-RVN® Insect Cell Line
Sf-RVN® Insect Cell Line has been adapted for growth in chemically defined medium to be used for the production of recombinant proteins, adeno-associated viruses (AAV) and viral-like particles (VLP).
- Product Information Sheet: EX-CELL® CD Insect Cell Medium
The EX-CELL® CD Insect Cell Medium is a specialized medium with a defined chemical composition, engineered to maximize the performance of insect cells. The formulation is animal-component free and has been formulated with L-Glutamine.
- White Paper: Development of a Novel Sf9 Rhabdovirus-Negative Cell Line (Sf-RVN®) and Companion Chemically Defined Medium
This whitepaper describes the development of a novel Sf9 Rhabdovirus-negative cell line, named Sf-RVN®, with companion chemically defined medium, the EXCELL® CD Insect Cell Medium. Combined, these two products form the Sf-RVN® Platform and offer a SfRhabdovirus free alternative for the BICS production of biologics.
- White Paper: Considerations for Bioreactor Process Development and Scale-up for Transient Transfection-based Lentivirus Production in Suspension
This white paper describes considerations for suspension-based, transient transfection bioreactor process development and scaleup of lentivirus production.
- Tool: CHOZN® Omics Explorer
- White Paper: A Multifaceted Strategy for Viral Safety in AAV Processes
This white paper describes an approach to adventitious viral risk mitigation which encompasses this multifaceted approach defined as “prevent, detect, remove”.
- Brochure: VirusExpress® 293 Production Platform
Draw on our experience to get on the fast track through production.
- Data Sheet: VirusExpress® AAV Production Platform
The VirusExpress® Platform offers a transfectionbased solution to Adeno-Associated Viral (AAV) production challenges, featuring a suspension adapted cell line, chemically defined medium, and a clinically relevant process dramatically reducing time to commercial production.
- Data Sheet: VirusExpress® Lentivirus Production Platform
The VirusExpress® Platform offers a transfection-based solution to lentiviral (LV) production challenges, featuring a suspension adapted cell line, chemically defined medium, and process with proven performance at 50 L scale.
CHOZN® Garis Sel untuk Produksi Biologis dan Protein Rekombinan
CHOZN® GS-/- Sel Line, Sel CHO yang Dimodifikasi dengan ZFN
Ditenagai oleh teknologiCompo Zr® Zinc Finger Nuclease (ZFN), garis selCHOZN® GS-/- yang terkenal dan banyak digunakan memungkinkan pengembangan garis sel yang cepat melalui seleksi metabolik tanpa penambahan MSX (inhibitor GS). Dengan stabilitas yang sangat baik, lebih dari 75% klon mempertahankan titer di atas 70% selama lebih dari 60 generasi.
Lini sel ini berasal dari ECACC CHO K1, yang telah diadaptasi untuk pertumbuhan suspensi dalam medium kimiawi yang didefinisikan dan bebas komponen hewan. Kinerja optimal dicapai ketika lini sel ini dipasangkan dengan medium kultur sel kimiawi yang didefinisikan dan suplemen.
Untuk informasi lebih lanjut tentang garis selCHOZN® GS-/-, silakan jelajahi di sini.
CHOZN® Elite Cell Line, Generasi Baru Host CHO untuk Meningkatkan Hasil Produksi Anda
CHOZN® Elite cell line adalah subklon dariCHOZN® GS-/- cellline yang unggul dalam efisiensi pengembangan sel (CLD), pertumbuhan yang dapat diprediksi, produksi protein, dan metrik kinerja lainnya dibandingkan dengan sel induknya. Sel ini sepenuhnya dapat dilacak dariCHOZN® GS-/- ZFN-modified CHO cell line dan telah disimpan sesuai standar cGMP, diuji karakterisasi, serta diuji terhadap agen adventisi untuk memenuhi persyaratan regulasi.
- Klone berproduksi tinggi: mencapai titer 7-13 g/L dengan vektor standar, menghasilkan penghematan biaya
- Stabilitas tinggi: 85% klon mempertahankan titer lebih dari 70% selama 60 generasi
- Profil pertumbuhan konsisten: 18 hingga 24 x10⁶ sel/mL untuk proses fed-batch
- Mudah diadopsi: plug-and-play dengan proses CLD yang sudah ada, untuk digunakan dalam proses fed-batch dan perfusi.
Untuk informasi lebih lanjut tentang garis selCHOZN® Elite, silakan jelajahi di sini.
Baris Sel untuk Produksi Protein Rekombinan, Partikel Mirip Virus, dan Vektor Virus
PlatformSf-RVN® untuk Meningkatkan Profil Keamanan Proses Bioteknologi Anda
PlatformSf-RVN® dengan garis sel serangga dan medium sel seranggaEX-CELL® CD menyediakan platform ber kinerja tinggi yang dioptimalkan untuk produksi protein rekombinan, vaksin virus seperti partikel serupa virus (VLP), dan vektor adeno-associated (AAV) untuk aplikasi terapi gen.
Sebagian besar sel Spodoptera frugiperda (seperti Sf-9) mengandung rhabdovirus yang dianggap sebagai kontaminan proses dan harus dihilangkan selama bioproses.
PlatformSf-RVN® merupakan sinergi antara garis sel Sf9 yang bebas rhabdovirus dan telah dipatenkan, yang menunjukkan karakteristik yang sama dengan sel Sf-9, dengan medium kimia yang didefinisikan secara khusus, yang diformulasikan untuk pertumbuhan dan produktivitas yang optimal.
Untuk informasi lebih lanjut tentang Garis Sel SeranggaSf-RVN®, silakan jelajahi di sini.
Platform Produksi LentivirusVirus Express®
PlatformVirus Express® Lentiviral adalah solusi berbasis transfeksi untuk mengatasi tantangan produksi lentivirus dan menawarkan proses hulu yang dapat diskalakan, dioptimalkan untuk produksi lentivirus berbasis suspensi. Platform ini menghemat waktu dalam pengembangan proses dan skalabilitas, serta dapat digunakan untuk mempercepat pengiriman terapi Anda kepada pasien.
Manfaat utama sel HEK293TVirus Express® meliputi:
- Baris sel tumbuh hingga kepadatan sel tinggi dan viabilitas dalam suspensi, serta memiliki waktu penggandaan yang rendah
- Dipilih untuk produksi lentivirus yang lebih tinggi menggunakan metode transfeksi sementara berbasis PEI
- Tersimpan di bank GMP, sepenuhnya terkaraterisasi, dan diuji terhadap agen adventisi
- Lisensi tersedia untuk penggunaan penelitian atau klinis/komersial.
- Sepenuhnya dapat dilacak dan dokumentasi tersedia untuk pengajuan regulasi
Untuk informasi tambahan tentang Platform LentivirusVirus Express®, silakan jelajahi di sini.
Platform ProduksiVirus Express® AAV
Platform ProduksiVirus Express® AAV menawarkan solusi berbasis transfeksi untuk tantangan produksi Virus Asosiasi Adeno (AAV), serta proses hulu yang dioptimalkan untuk produksi AAV berskala besar berbasis suspensi. Platform ini menghemat waktu dalam pengembangan proses dan skalabilitas, serta dapat digunakan untuk mempercepat penyediaan terapi Anda kepada pasien.
Manfaat utama sel HEK293Virus Express® meliputi:
- Baris sel tumbuh hingga kepadatan sel tinggi dan viabilitas dalam suspensi, serta memiliki waktu penggandaan yang rendah
- Dipilih untuk produksi AAV yang lebih tinggi menggunakan metode transfeksi sementara berbasis PEI
- Tersimpan di bank GMP, sepenuhnya terkaraterisasi, dan diuji terhadap agen adventisi
- Lisensi tersedia untuk penggunaan penelitian atau klinis/komersial
- Dapat dilacak sepenuhnya dan dokumen tersedia untuk pengajuan regulasi